Learn More
Novus Biologicals™ Cannabinoid R1/CB1/CNR1 Knockout HeLa Cell Lysate
This product is for research use only and is not approved for use in humans or in clinical diagnosis.
Manufacturer:  Novus Biologicals™ NBP266170
Catalog No. NBP266170
Add to cart
Description
Description
The Cannabinoid R1/CB1/CNR1 Knockout HeLa Cell Lysate is derived from human. It has been validated for the following applications: Western Blot.
Specifications
Specifications
LYOPH
Human
0.1 mg cell homogenate lyophilized in RIPA buffer made with double-knockout cell lines
Cannabinoid R1/CB1/CNR1
Western Blot
2 Vials
Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.