Learn More
Cannabinoid R1/CB1/CNR1 Mouse anti-Human, Alexa Fluor 488, Clone: 368302, R&D Systems™
Mouse Monoclonal Antibody
Manufacturer:  R&D Systems FAB3834G100UG
Catalog No. FAB3834G100
Add to cart
This item is not returnable. View return policy
Description
Description
Cannabinoid R1/CB1/CNR1 Monoclonal specifically detects Cannabinoid R1/CB1/CNR1 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
Cannabinoid R1/CB1/CNR1
Monoclonal
0.2 mg/mL
Flow Cytometry 0.25-1 ug/10^6 cells
NP_057167
CNR1
HEK293 human embryonic kidney cell line transfected with human Cannabinoid R1/CB1/CNR1 Met1-Leu472 Accession # NP_057167
100 ug
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Monoclonal
Stains Cannabinoid R1/CB1/CNR1-transfected cells but not irrelevant transfectants.
Flow Cytometry
368302
Alexa Fluor 488
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
CANN6, CANN6CB1R, cannabinoid receptor 1 (brain), CannabinoidR1, CB1, CB1A, CB1K5, CB1R, CB-R, CB-RCB1cannabinoid receptor 1, central cannabinoid receptor, CNR, CNR1, SKR6R
Mouse
IgG2a
RUO
Primary
1268
Human
SDS
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)