Learn More
Cannabinoid R2/CB2/CNR2 Mouse anti-Human, Alexa Fluor™ 750, Clone: 352110R, R&D Systems™
Mouse Monoclonal Antibody
Manufacturer:  R&D Systems FAB36551RS100UG
Catalog No. FAB36551RS1
Add to cart
This item is not returnable. View return policy
Description
Description
Cannabinoid R2/CB2/CNR2 Monoclonal specifically detects Cannabinoid R2/CB2/CNR2 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
Cannabinoid R2/CB2/CNR2
352110R
Flow Cytometry 0.25-1 μg/106 cells
NP_001832
cannabinoid receptor 2 (macrophage), CannabinoidR2, CB-2, CB2cannabinoid receptor 2, CB2R, CNR2, CX5, CX5testis-dominant CNR2 isoform CB2, hCB2
Human embryonic kidney cell line HEK293-derived transfected with human Cannabinoid R2/CB2/CNR2, Met1-Cys360, Accession # NP_001832
Protein A or G purified from cell culture supernatant
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Monoclonal
Detects human Cannabinoid R2/CB2/CNR2 in direct ELISAs.
Flow Cytometry
Alexa Fluor 750
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Purified, Supernatant, Conjugated
Mouse
IgG2b
100 μg
Primary
1269
Human