Learn More
Sklar InstrumentsSupplier Diversity Partner Cannula Instrument Cleaning Brush, Sklar, 15 in., Nylon / Stainless Steel. This product is 9mm x 15 inches. Pack of 12. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 101665
Cannula Instrument Cleaning Brush, Sklar, 15 in., Nylon / Stainless Steel. Cannula Instrument Cleaning Brushes are used in the multiple-step process of sterilizing a reusable cannula. Following soaking in enzymatic solution, the cannula should be cleaned manually with a brush and detergent or ultrasonically. This product is 9mm x 15 inches. Pack of 12.
Catalog No. 50-143-7077