Learn More
Sklar InstrumentsSupplier Diversity Partner Cannula Instrument Cleaning Brush, Sklar, 25mm x 18 in. Cannula Instrument Cleaning Brush Nylon Bristle End, Nylon / Stainless Steel. PK/3. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 101369
Cannula Instrument Cleaning Brush, Sklar, 25mm x 18 in. Cannula Instrument Cleaning Brush Nylon Bristle End, Nylon / Stainless Steel. Sklar's Cannula Instrument Cleaning Brushes are used in the multiple-step process of sterilizing a reusable cannula. Following soaking in enzymatic soluntion the cannula should be cleaned manually with a brush and detergent or ultrasonically. Sklar cannula cleaning brushes are designed for manual cleaning and are available in several sizes in order to properly decontaminate specific size cannulas used for specific procedures. This product has a 25mm wide cleaning brush and an overall length of 18 inches. It also features a loop at the end of the handle for improved user grip and control.
Catalog No. 50-143-6965