Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CANT1 Mouse anti-Human, Clone: OTI7F8, lyophilized, TrueMAB
Mouse Monoclonal Antibody
Manufacturer: OriGene CF805629
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This protein encoded by this gene belongs to the apyrase family. It functions as a calcium-dependent nucleotidase with a preference for UDP. Mutations in this gene are associated with Desbuquois dysplasia with hand anomalies.Specifications
CANT1 | |
Monoclonal | |
Unconjugated | |
CANT1 | |
Lyophilized | |
CANT1 | |
Human recombit protein fragment corresponding to amino acids 63-401 of human CANT1 produced in E.coli. | |
Affinity Chromatography | |
RUO | |
Antibody | |
Monoclonal | |
Human |
Immunohistochemistry (Paraffin), Western Blot | |
OTI7F8 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q8WVQ1 | |
DBQD, DBQD1, EDM7, SCAN-1, SCAN1, SHAPY | |
Mouse | |
IgG1 | |
100 μg | |
-20° C, Avoid Freeze/Thaw Cycles | |
Primary | |
124583 |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title