Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Capecitabine, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
50mg
This item is not returnable. View return policy
154361-50-9
C15H22FN3O6
359.354
GAGWJHPBXLXJQN-UORFTKCHSA-N
capecitabine,xeloda,capiibine,capecitibine,caxeta,xabine,capecitabine usan,capecitabina,capecitabinum,capecitabin
60953
CHEBI:31348
pentyl N-[1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-methyloxolan-2-yl]-5-fluoro-2-oxopyrimidin-4-yl]carbamate
CCCCCOC(=O)NC1=NC(=O)N(C=C1F)C2C(C(C(O2)C)O)O
This item is not returnable. View return policy
154361-50-9
C15H22FN3O6
359.354
GAGWJHPBXLXJQN-UORFTKCHSA-N
capecitabine,xeloda,capiibine,capecitibine,caxeta,xabine,capecitabine usan,capecitabina,capecitabinum,capecitabin
60953
CHEBI:31348
pentyl N-[1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-methyloxolan-2-yl]-5-fluoro-2-oxopyrimidin-4-yl]carbamate
CCCCCOC(=O)NC1=NC(=O)N(C=C1F)C2C(C(C(O2)C)O)O

Prodrug of 5-Fluorouracil (Cat. No. 3257). Inhibits DNA synthesis

Prodrug of 5-Fluorouracil (5-FU) (Cat. No. 3257). Selectively activated in tumor cells by thymidine phosphorylase; inhibits DNA synthesis upon conversion to 5-FU. Orally available.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 50mg
Formula Weight 359.35
Percent Purity >98%
Chemical Name or Material Capecitabine

Recommended Storage : Store at +4°C