Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CapG Rabbit anti-Human, DyLight 350, Polyclonal, Novus Biologicals
Rabbit Polyclonal Antibody
Manufacturer: Novus Biologicals NBP300258UV
Description
CapG Polyclonal antibody specifically detects CapG in Human samples. It is validated for Western Blot, ELISASpecifications
CapG | |
DyLight 350 | |
50mM Sodium Borate | |
actin-regulatory protein CAP-G, AFCP, capping protein (actin filament), gelsolin-like, gelsolin-like capping protein, macrophage capping protein, macrophage-capping protein, MCPActin regulatory protein CAP-G | |
Produced in rabbits immunized with purified, recombinant CapG (AAH14549.1; Met1-Lys348). | |
Affinity purified | |
RUO | |
Store at 4°C in the dark. | |
Polyclonal | |
Human |
ELISA, Western Blot | |
Western Blot, ELISA | |
Purified | |
Rabbit | |
IgG | |
0.1 mL | |
Cell Biology, Cellular Markers | |
Primary | |
822 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title