Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Capicua Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 2 publications
$270.00 - $566.00
Specifications
| Antigen | Capicua |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000-1:2500, Immunocytochemistry/Immunofluorescence 1:50, Immunoprecipitation 2-5 ug/mg lysate, Knockout Validated |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, KnockDown |
| Classification | Polyclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB11059906T
![]() |
Novus Biologicals
NB11059906SS |
0.025 mg |
Each for $270.00
|
|
|||||
NB11059906
![]() |
Novus Biologicals
NB11059906 |
0.1 mg |
Each for $566.00
|
|
|||||
Description
Capicua Polyclonal specifically detects Capicua in Human, Mouse, Rat, Canine samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Knockout Validated.Specifications
| Capicua | |
| Western Blot 1:1000-1:2500, Immunocytochemistry/Immunofluorescence 1:50, Immunoprecipitation 2-5 ug/mg lysate, Knockout Validated | |
| Polyclonal | |
| Rabbit | |
| Cancer, Neuroscience | |
| PBS with 0.02% Sodium Azide | |
| capicua homolog (Drosophila), KIAA0306capicua (Drosophila) homolog, protein capicua homolog | |
| CIC | |
| IgG | |
| Affinity Purified | |
| 250 kDa |
| 1 mg/mL | |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, KnockDown | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat, Canine | |
| Q96RK0 | |
| 23152 | |
| The epitope recognized by this antibody maps to a C-terminal region within residue 1500-1608 of human Capicua. [NP_055940.3] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title