Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Capicua Antibody, DyLight 550, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB11059906R
Description
Capicua Polyclonal specifically detects Capicua in Human, Mouse, Rat, Canine samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Knockout Validated.Specifications
| Capicua | |
| Polyclonal | |
| Western Blot, Immunocytochemistry/Immunofluorescence, Knockout Validated | |
| Q96RK0 | |
| CIC | |
| The epitope recognized by this antibody maps to a C-terminal region within residue 1500-1608 of human Capicua. [NP_055940.3] | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Canine | |
| IgG |
| Western Blot, Immunocytochemistry, Immunofluorescence, KnockDown | |
| DyLight 550 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| capicua homolog (Drosophila), KIAA0306capicua (Drosophila) homolog, protein capicua homolog | |
| Rabbit | |
| 250 kDa | |
| 0.1 mL | |
| Cancer, Neuroscience | |
| 23152 | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction