DBI/Sala™ Delta II Construction Vest-Style Harnesses

No-Tangle™ design provides high level of comfort and easy donning

Manufacturer: DBI Sala Inc. 1103512

 View more versions of this product

Catalog No. 18-706

  Check Availability
Add to Cart

    Specifications

    Product Type Construction Vest Style Harness
    Style Vest
    D-Ring Location Back, Side
    Webbing Material Polyester
    Capacity (English) 420 lb.
    Capacity (Metric) 190.5kg
    Includes 190.5kg
    Buckle Style (Leg) Pass through
    Buckle Style (Shoulder) Parachute
    Buckle Style (Chest) Quick connect
    Certifications/Compliance ANSI, OSHA
    Color Yellow and Black
    For Use With (Application) Construction
    Size Universal

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit