Learn More
3M™ DBI-Sala™ ExoFit™ XP Vest-Style Full Body Harnesses
The design incorporates a single piece of material that wraps around you in the shape of an X, for the ultimate in no-tangle donning and comfortable security. Its 3-D mesh lining allows moisture to evaporate and cool air to circulate throughout the padding, which is removable and washable.
Supplier: 3M™ 1110103
Description
- Single piece of material that wraps around in shape of X
- State-of-the-art 3-D mesh lining allows moisture to evaporate and cool air to circulate throughout the shoulder, back, hip and leg pads
- Cushioned leg and hip straps offers comfort and additional security during fall
- Soft edging prevents chafing and lightweight webbing is strong without feeling stiff and coarse
- Spring loaded “stand-up” adjustable dorsal D-ring ensures easy connection to lanyard/SRL without straining to reach it
- Five-point adjustment system with built-in impact indicator on dorsal D-ring
- Color-coded quick-connect buckles (silver leg buckles and black chest buckle) makes donning easy
- “Easy-grip” end tabs provides hold onto when adjusting the straps
- Removable and washable padding
- Ultimate in no-tangle donning and comfortable security
Specifications
X-Large | |
Vest |