Learn More
3M™ DBI-Sala™ Ultra-Lok™ Web Self-Retracting Lifelines
Consists of anti-ratcheting twin disc brake system with an audio indicator for security
Manufacturer:  3M™ 3103213
 View more versions of this product
Catalog No. 19-809-363
Add to cart
Description
Description
  • Comes with durable glass-filled polyurethane housing, 1 in. web lifeline and aluminum side plates for maximum durability and minimum weight
  • Corrosion resistant stainless-steel working components
  • 11 ft. 1 in. nylon web lifeline for reduced weight
  • Reserve lifeline retention system for added safety
  • Self-locking swivel snap hook with impact indicator
  • Swiveling anchorage loop to prevent twisting of lifeline
Compliance
Meets or exceeds all applicable industry standards including OSHA, ANSI and the stringent ANSI Z359
Specifications
Specifications
Self retracting lifeline
OSHA, ANSI, ANSI Z359
6.1m
2.54cm
2.49kg
Carbiner, anti ratcheting twin disk brake
20 ft.
1 in.
5.5 lb.