Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3M™ DBI-Sala™ Talon™ Self-Retracting Lifelines
Engineered for reliable protection with automatic lock and unlock system
Supplier: 3M™ 3101009
Description
- Designed for smooth consistent operation
- Lightweight design (3 lb.) and quick-connect system gives freedom of movement without compromising safety
- Due to small size it can be connected directly to back of harness for added mobility
- Compact form with order picker handle
- All metal braking system for durability
- Self-locking swivel snap hook with impact indicator
Compliance
Meets or exceeds all applicable industry standards including OSHA, ANSI and stringent ANSI Z359
Specifications
Self-retracting lifeline | |
OSHA, ANSI, ANSI Z359 | |
8 ft. | |
4 kN |
2.43 m | |
Cab mount bracket, self locking swivel snap hook | |
900 lb. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction