Learn More
3M™ DBI-Sala™ Talon™ Self-Retracting Lifelines
Engineered for reliable protection with automatic lock and unlock system
Manufacturer:  3M™ 3101008
 View more versions of this product
Catalog No. 19-066-834
Add to cart
Description
Description
  • Designed for smooth consistent operation
  • Lightweight design (3 lb.) and quick-connect system gives freedom of movement without compromising safety
  • Due to small size it can be connected directly to back of harness for added mobility
  • Compact form with order picker handle
  • All metal braking system for durability
  • Self-locking swivel snap hook with impact indicator
Compliance
Meets or exceeds all applicable industry standards including OSHA, ANSI and stringent ANSI Z359
Specifications
Specifications
Safety
Cab mount bracket, snap locking hook
900 lb.
8 ft.
Self retracting lifeline
OSHA, ANSI, ANSI Z359
4kN
2.43m