Learn More
Capital Safety™ DBI-Sala™ Talon™ Self-Retracting Lifelines
Engineered for reliable protection with automatic lock and unlock system
Manufacturer:  Capital Safety™ 3101010X
 View more versions of this product
Catalog No. 19-120-3584
Add to cart
Description
Description
  • Designed for smooth consistent operation
  • Lightweight design (3 lb.) and quick-connect system gives freedom of movement without compromising safety
  • Due to small size it can be connected directly to back of harness for added mobility
  • Compact form with order picker handle
  • All metal braking system for durability
  • Self-locking swivel snap hook with impact indicator
Compliance
Meets or exceeds all applicable industry standards including OSHA, ANSI and stringent ANSI Z359
Specifications
Specifications
Self retracting lifeline
900 lb.
8 ft.
Cab mount bracket, Steel carabiner
4kN
2.43m