Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Capitol Vial Collection Cups

Thermo Scientific™ Capitol Vial Collection Cups are used for for traditional and on-site testing.

Manufacturer:  Thermo Scientific™ 10126

 View more versions of this product

Catalog No. 14-377-838


Add to cart

Description

Description

Thermo Scientific™ Capitol Vial Collection Cups are used for traditional and on-site testing. Choose from two different beaker-style collection cups.

  • Pour spout present for easy transfer of sample into smaller containers
  • With or without temperature indicator strips
Specifications

Specifications

236 mL
Capitol Vial Collection Cup
8 oz.
Collection Cup with temperature strip; 8 oz. (236 mL)
Plastic
236 mL
Natural
Temperature Strip
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.