Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Capitol Vial EPA Vials

Patented flip top with permanent hinge lid

Manufacturer:  Thermo Scientific™ 04HP83HCNP

 View more versions of this product

Catalog No. 14-377-810


Add to cart

Description

Description

  • Capacity, 118mL (4 oz.)
  • 100mL EPA fill line
  • High-clarity resin
  • With or without sodium thiosulfate pill, string

Specifications

Specifications

118 mL
Used to test for coliform
EPA Vial
Clear
Flip-Top
Vial and String
3.99 oz.
Resin
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit