Learn More
Thermo Scientific™ Capitol Vial Flip-Top 2.0 oz. Vials
Thermo Scientific™ Capitol Vial Flip-Top 2.0 oz. Vials provide economical storage and packaging of a wide range of liquids and solids.
$137.00 - $139.50
Specifications
Diameter (English) Inner 0.9 in.
Diameter (Metric) Inner 23 mm
Length (Metric) 84 mm
Material Polypropylene
Length (English) 3.3 in.
View More Specs
Products 13
Catalog Number Mfr. No. Color Price Quantity & Availability  
Catalog Number Mfr. No. Color Price Quantity & Availability  
14-378-11
View Documents
Thermo Scientific™
02 BLACK
Black Case of 600 for $139.50
Add to cart
14-378-13
View Documents
Thermo Scientific™
02 DKORANGE
Dark Orange Case of 600 for $139.50
Add to cart
14-377-96
View Documents
Thermo Scientific™
02 EVERGREEN
Evergreen Case of 600 for $139.50
Add to cart
14-377-97
View Documents
Thermo Scientific™
02 FREGORED
Frego Red Case of 600 for $139.50
Add to cart
14-377-98
View Documents
Thermo Scientific™
02 GOLD
Gold Case of 600 for $139.50
Add to cart
14-377-99
View Documents
Thermo Scientific™
02 LIME
Lime Case of 600 for $139.50
Add to cart
1437803
View Documents
Thermo Scientific™
02 PINK
Pink Case of 600 for $139.50
Add to cart
14-378-05
View Documents
Thermo Scientific™
02 ROYALBLUE
Royal Blue Case of 600 for $139.50
Add to cart
14-378-07
View Documents
Thermo Scientific™
02 SKYBLUE
Sky Blue Case of 600 for $139.50
Add to cart
14-377-90
View Documents
Thermo Scientific™
02 YELLOW
Yellow Case of 600 for $139.50
Add to cart
14-378-08
View Documents
Thermo Scientific™
02 TURQ
Turquoise Case of 600 for $139.50
Add to cart
14-378-04
View Documents
Thermo Scientific™
02 PURPLE
Purple Case of 600 for $139.50
Add to cart
14-377-88 Edge
View Documents
Thermo Scientific™
02CL
Natural Case of 600 for $137.00
Edge
Add to cart
Description
Description
Thermo Scientific™ Capitol Vial Flip-Top 2.0 oz. Vials provide economical storage and packaging of a wide range of liquids and solids.
  • Aseptic, airtight and leakproof construction
  • 100% FDA-approved food-grade polypropylene vials
  • Permanent hinge lid allows easy one-handed opening of vial
  • Shoulderless, liner-less containers
  • 0.9 I.D. x 3.3in.L (23 x 84mm)
  • Available in 22 different colors
Ordering Information:
Case of 600
Recommended for:
Biomaterial storage, biopsy transport, DNA storage, dairy testing, diagnostic product packaging, environmental sampling, water testing
Warranty: 90 days
Specifications
Specifications
0.9 in.
84 mm
3.3 in.
23 mm
Polypropylene
Sterile
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.