Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Capitol Vial Flip-Top Vials with Lock Seal

Thermo Scientific™ Capitol Vial Flip-Top Vials with Lock Seal are similar to standard flip-top vials except they contain an additional lock-seal tab.

Manufacturer:  Thermo Scientific™ 08LS

 View more versions of this product

Catalog No. 14-377-818


Add to cart

Description

Description

Thermo Scientific™ Capitol Vial Flip-Top Vials with Lock Seal are similar to standard flip-top vials except they contain an additional lock-seal tab.

  • 100% FDA approved food grade polypropylene
  • Sterile, airtight and leak-resistant construction
  • Permanent hinged lid allows easy one-handed opening of vial
  • Shoulderless, linerless containers
  • Available in capacities ranging from 2 to 10 oz. (59 to 296mL)

Recommended for:

Biomaterial storage, biopsy transport, DNA storage, dairy testing, diagnostic product packaging, environmental sampling, water testing

Specifications

Specifications

Sterile
8 oz.
Polypropylene
236 mL
Natural
8 oz.; Natural
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.