Learn More
Thermo Scientific™ Capitol Vial Flip-Top Vials with Lock Seal
Thermo Scientific™ Capitol Vial Flip-Top Vials with Lock Seal are similar to standard flip-top vials except they contain an additional lock-seal tab.
$49.83 - $238.00
Specifications
Sterility Sterile
View More Specs
Products 10
Catalog Number Mfr. No. Volume (English) Material Color Price Quantity & Availability  
Catalog Number Mfr. No. Volume (English) Material Color Price Quantity & Availability  
14-377-89
View Documents
Thermo Scientific™
02LS
1.5 oz. Polypropylene Natural
Case of 600 for $238.00
Add to cart
14-377-20
View Documents
Thermo Scientific™
04HPLS
4 oz. High Profile Polypropylene Natural
Case of 250 for $94.90
Add to cart
14-377-813
View Documents
Thermo Scientific™
04HPLS ROYALBLUE
4 oz. Polypropylene Royal Blue
Case of 250 for $94.40
Add to cart
14-377-915
View Documents
Thermo Scientific™
04LPLS
4 oz. Polypropylene Natural
Case of 250 for $106.10
Add to cart
14-377-855
View Documents
Thermo Scientific™
04LPLSBLACK
4 oz. Polypropylene Black
Case of 250 for $133.70
Add to cart
14-377-858
View Documents
Thermo Scientific™
04LPLSYELLOW
4 oz. Polypropylene Yellow
Case of 250 for $137.70
Add to cart
14-377-818
View Documents
Thermo Scientific™
08LS
8 oz. Polypropylene Natural
Case of 140 for $84.00
Add to cart
14-377-820
View Documents
Thermo Scientific™
10LSCL
10 oz. Polypropylene Natural
Case of 100 for $64.00
Add to cart
22-039-069
View Documents
Thermo Scientific™
10LSWHITE
10 oz. Polypropylene White
Case of 100 for $49.83
Add to cart
22-039-070
View Documents
Thermo Scientific™
10LSBLACK
10 oz. Polypropylene Black
Case of 100 for $49.83
Add to cart
Description
Description
Thermo Scientific™ Capitol Vial Flip-Top Vials with Lock Seal are similar to standard flip-top vials except they contain an additional lock-seal tab.
  • 100% FDA approved food grade polypropylene
  • Sterile, airtight and leak-resistant construction
  • Permanent hinged lid allows easy one-handed opening of vial
  • Shoulderless, linerless containers
  • Available in capacities ranging from 1.5 to 10 oz. (44.3 to 296mL)
Recommended for:
Biomaterial storage, biopsy transport, DNA storage, dairy testing, diagnostic product packaging, environmental sampling, water testing
Warranty: 90 days
Specifications
Specifications
Sterile
SDS
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.