Learn More
Greiner Bio-One CapMat, for use with 96-well Masterblock™ Microplates
Sealers for microplates
Manufacturer:  Greiner Bio-One 381070
Catalog No. 07-000-176
Add to cart
Description
Description
  • For use with 96-well Masterblock Microplates
Specifications
Specifications
Not Autoclavable
Free of detectable DNase/RNase, human DNA and pyrogens; resistant to DMSO; non-cytotoxic
Non-sterile
50
No
-60°C to +65°C
1mL and 0.5mL 96-well MASTERBLOCK, standard 96-well polypropylene microplates and 96-well F-, U-, and μClear-bottom polystyrene microplates
Translucent
EVA
96
1
No
Round