Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CAPN1 Mouse anti-Human, Clone: 1376CT809.24.85, Abnova™

Mouse monoclonal antibody raised against synthetic peptide of human CAPN1.

Manufacturer:  Abnova Corporation MAB12351

Catalog No. 89-933-720


Add to cart

Description

Description

The calpains, calcium-activated neutral proteases, are nonlysosomal, intracellular cysteine proteases. The mammalian calpains include ubiquitous, stomach-specific, and muscle-specific proteins. The ubiquitous enzymes consist of heterodimers with distinct large, catalytic subunits associated with a common small, regulatory subunit. This gene encodes the large subunit of the ubiquitous enzyme, calpain 1. [provided by RefSeq]

Specifications

Specifications

CAPN1
1376CT809.24.85
Mouse monoclonal antibody raised against synthetic peptide of human CAPN1.
In PBS (0.09% sodium azide)
CANP/CANP1/CANPL1/muCANP/muCL
Mouse
IgG1, kappa
400 uL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
Flow Cytometry, Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
Flow Cytometry (1:25) Immunohistochemistry (1:25) Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Liquid
CAPN1
A synthetic peptide (conjugated with KLH) corresponding to human CAPN1.
Protein G purification
RUO
Primary
823
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit