Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CAPN1 Rabbit anti-Human, Unconjugated, Clone: ABFI-3, Abnova™
Rabbit Monoclonal Antibody.
Supplier: Abnova Corporation MAB19688
Description
CAPN1 Monoclonal antibody specifically detects CAPN1 in Human samples. It is validated for ICC, IHC, WB-Ti, IF, Flow Cyt.Specifications
CAPN1 | |
Monoclonal | |
Unconjugated | |
CAPN1 | |
Rabbit | |
100 μg | |
823 | |
Store at -20°C for one year. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for a longer time. Avoid repeated freezing and thawing. | |
IgG |
Immunocytochemistry, Immunohistochemistry, Western Blot, Immunofluorescence, Flow Cytometry | |
ABFI-3 | |
calpain 1, (mu/I) large subunit | |
CANP, CANP1, CANPL1, muCANP, muCL | |
A synthetic peptide corresponding to human CAPN1 | |
Primary | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction