Learn More
Aviva Systems Biology Corp CAPN10 Antibody - N-terminal region (ARP52404_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP52404_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CAPN10. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CAPN10. Alternate names: CANP10, NIDDM1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-60728