Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Calpain 12 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA549994
Description
The antibody detects endogenous levels of total CAPN12 protein.
Calpain 12 Polyclonal antibody specifically detects Calpain 12 in Human, Mouse samples. It is validated for Western Blot.Specifications
Calpain 12 | |
Polyclonal | |
Unconjugated | |
CAPN12 | |
calcium-activated neutral proteinase 12; calpain 12; calpain-12; CANP 12; CAPN12 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
147968, 60594 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q6ZSI9, Q9ER56 | |
CAPN12 | |
Synthesized peptide derived from internal of human CAPN12. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction