Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CAPNS2 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549194

Catalog No. PA549194

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Calcium-regulated non-lysosomal thiol-protease which catalyzes limited proteolysis of substrates involved in cytoskeletal remodeling and signal transduction. This small subunit may act as a tissue-specific chaperone of the large subunit, possibly by helping it fold into its correct conformation for activity.
Specifications

Specifications

CAPNS2
Polyclonal
PBS with 0.09% sodium azide
Q96L46, Q9D7J7
Calpain small subunit 2, CSS2, Calcium-dependent protease small subunit 2, CAPNS2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
69543, 84290
Western Blot
Unconjugated
CAPNS2
Liquid
CAPNS2
KLH conjugated synthetic peptide between 55-83 amino acids from the Central region of human CAPNS2.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit