Learn More
Honeywell Burdick & Jackson™ Capping Reagent, 84/16 THF/NMI (v/v)
BioSyn™, for DNA/RNA synthesis
Supplier:  Honeywell Burdick & Jackson™ BR65010
 View more versions of this product
Catalog No. 60-048-383
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
This item is not returnable. View return policy
Description
Description
Burdick & Jackson™ BioSyn™ reagents & solvents for DNA/RNA synthesis
  • Honeywell Burdick & Jackson™ BioSyn™ Solvents and Reagents are specifically processed and purified to exacting specifications, ensuring outstanding synthesis efficiency
  • ideal for oligonucleotide synthesis regardless of the application
  • All solvents and reagents are filtered (0.2-micron for oxidation reagents and 0.1-micron for all other reagents) to minimize particulate contamination
Specifications
Specifications
616-47-7
0.910 g/cm3
HDPE NOWPAK
Room Temperature
Liquid
2 x 10 L