Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ε-Caprolactone monomer, 99%, ACROS Organics™

Click to view available options
Quantity:
250g
1kg
2.5kg
10kg
Packaging:
Glass bottle
Plastic drum
502-44-3
C6H10O2
114.144
MFCD00003267
PAPBSGBWRJIAAV-UHFFFAOYSA-N
6-hexanolactone,epsilon-caprolactone,2-oxepanone,caprolactone,hexan-6-olide,6-hexanolide,hexano-6-lactone,1,6-hexanolide,e-caprolactone,1-oxa-2-oxocycloheptane
10401
CHEBI:17915
oxepan-2-one
C1CCC(=O)OCC1
502-44-3
C6H10O2
114.144
MFCD00003267
PAPBSGBWRJIAAV-UHFFFAOYSA-N
6-hexanolactone,epsilon-caprolactone,2-oxepanone,caprolactone,hexan-6-olide,6-hexanolide,hexano-6-lactone,1,6-hexanolide,e-caprolactone,1-oxa-2-oxocycloheptane
10401
CHEBI:17915
oxepan-2-one
C1CCC(=O)OCC1
Refractive Index 1.4620 to 1.4640 (20°C, 589nm)
Quantity 2.5kg
Infrared Spectrum Authentic
Color Colorless
Water 0.05% max.
Packaging Glass bottle
Density 1.0300g/mL
Assay Percent Range 99%
Formula Weight 114.14
Physical Form Liquid
Percent Purity ≥98.5%
Chemical Name or Material ε-Caprolactone monomer
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.