Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ CAPS, 0.5M buffer soln., pH 9.0

Click to view available options
Quantity:
250mL
500mL
1135-40-6
C9H19NO3S
221.315
MFCD00003837
PJWWRFATQTVXHA-UHFFFAOYSA-N
70815
3-(cyclohexylamino)propane-1-sulfonic acid
C1CCC(CC1)NCCCS(=O)(=O)O
1135-40-6
C9H19NO3S
221.315
MFCD00003837
PJWWRFATQTVXHA-UHFFFAOYSA-N
70815
3-(cyclohexylamino)propane-1-sulfonic acid
C1CCC(CC1)NCCCS(=O)(=O)O
Concentration 0.5M buffer soln.
Physical Form Liquid
pH 9.0
Quantity 250mL
Chemical Name or Material CAPS buffer soln.