Learn More
DWK Life Sciences DURAN™ Caps and Inserts for DURAN™ Laboratory Glass Bottles
Manufacturer:  DWK Life Sciences 1088655
 View more versions of this product
Catalog No. 09-841-268
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Typical applications: storage or transport of gas generating media, autoclaving of media.
Specifications
Specifications
54mm
Polypropylene with welded in PTFE Membrane
Membrane Screw Cap
Duran™ laboratory glass bottles
GL45
Yes
Blue
For storage or transport of gas generating media, autoclaving of media
25mm
Caps and Inserts