Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
DWK Life Sciences DURAN™ Caps and Inserts for DURAN™ Laboratory Glass Bottles
Specifications
Autoclavable Autoclavable
View More Specs
Products 6
Catalog Number Mfr. No. Color Material Type Closure Size Price Quantity & Availability  
Catalog Number Mfr. No. Color Material Type Closure Size Price Quantity & Availability  
09-841-112
View Documents Promotion Details
DWK Life Sciences
1129814
Blue Polypropylene Screw Cap, Caps and Inserts GL14
Pack of 2 for $28.55
N/A
09-841-113
View Documents Promotion Details
DWK Life Sciences
1156292
Red Polybutylene Screw Cap, Caps and Inserts GL14
Pack of 2 for $15.89
N/A
09-841-116
View Documents Promotion Details
DWK Life Sciences
1129815
White PTFE, Silicone Insert, Insert, 1.6mm GL14
Pack for $46.35
N/A
09-841-117
View Documents Promotion Details
DWK Life Sciences
1129816
White PTFE, Silicone Insert, Insert, 3mm GL14
Case for $57.40
N/A
09-841-118
View Documents Promotion Details
DWK Life Sciences
1129817
White PTFE, Silicone Insert, Insert, 3.2mm GL14
Pack for $46.35
N/A
09-841-119
View Documents Promotion Details
DWK Life Sciences
1129818
White PTFE, Silicone Insert, Insert, 6mm GL14
Pack for $46.35
N/A
Description
Description
Flexible modular system with four different tubing diameters (1.6 mm; 3.0 mm; 3.2 mm and 6.0 mm) can be connected. Sterile pressure equalization is possible through use of syringe filter. Unused ports can be provided with a blind cap.
Caps and inserts for use with DURAN Laboratory Glass Bottles.
Specifications
Specifications
Autoclavable