Thermo Scientific™ Caps for Nunclon™ Delta Flasks
Protect your valuable cell culture samples. These caps for Thermo Scientific™ Nunclon™ Delta Flasks are offered in two styles—vent/close, filter—to meet application needs.
Manufacturer: Thermo Scientific™ 158892
 View more versions of this product
Catalog No. 12-565-622
  Check Availability
Add to Cart
Disclaimers:
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
    Specifications
    Closure Type Vent/Close
    Material HDPE
    For Use With 25cm2 EasY Flask
    Closure Vent/Close
    Closure Color Blue
    Description Sterile, HDPE closure
    No. per Pack 1
    No. per Case 100
    Protect your valuable cell culture samples. These caps for Thermo Scientific™ Nunclon™ Delta Flasks are offered in two styles—vent/close, filter—to meet your application needs.
    Vent/close Caps
    • EasY Flask vent/close caps feature a "Y" mark that allows visual verification
    • Vent position easily distinguised even what flasks are stacked in incubators
    Filter Caps
    • Filter membrane provides effective barrier against microorganisms
    • Well-defined, hydrophobic filter membrane offers continuous venting with consistent gas exchange and no lot-to-lot variation
    • Chemically robust membrane meets USP class VI standards and is produced without animal-derived substances
    Certifications:
    • Sterilized by gamma irradiation and certified to be nonpyrogenic
    Ordering Information:
    • Case of 100
  • Description & Specifications
Related Products
Product Content Feedback