Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Capsaicin, Natural, Spectrum™

Click to view available options
Quantity:
1 g
5 g
100 g
1 kg
Packaging:
Amber Glass Bottle
404-86-4
C18H27NO3
305.42
YKPUWZUDDOIDPM-SOFGYWHQSA-N
(6E)-N-[(4-hydroxy-3-methoxyphenyl)methyl]-8-methylnon-6-enamide
COC1=CC(CNC(=O)CCCC\C=C\C(C)C)=CC=C1O
404-86-4
C18H27NO3
305.42
YKPUWZUDDOIDPM-SOFGYWHQSA-N
(6E)-N-[(4-hydroxy-3-methoxyphenyl)methyl]-8-methylnon-6-enamide
COC1=CC(CNC(=O)CCCC\C=C\C(C)C)=CC=C1O

C1414, 404-86-4, C18H27NO3

Spectrum™ Capsaicin, Natural is an irritant that produces a burning sensation when it comes into contact with tissue. It is the active component of the genus Capsicum plant and belong to chili pepper family. In its pure form it is a waxy to crystalline compound that is colorless, odorless, volatile and hydrophobic. It is commonly used in food products as an additive to give a spicy or "heat" taste sensation. In the medical realm it is used as an analgesic in nasal spays, topical ointments and in dermal patches to relieve pain. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
Packaging Amber Glass Bottle
Ignition Residue 0.01
Loss on Drying 0.01
Quantity 1 kg
Melting Range 57-66°C
CAS Max % 1
UN Number UN2811