Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Capsaicin, USP, Spectrum™

Click to view available options
Quantity:
1 g
5 g
25 g
100 g
1 kg
Packaging:
Amber Glass Bottle
404-86-4
C18H27NO3
305.42
YKPUWZUDDOIDPM-SOFGYWHQSA-N
(6E)-N-[(4-hydroxy-3-methoxyphenyl)methyl]-8-methylnon-6-enamide
COC1=CC(CNC(=O)CCCC\C=C\C(C)C)=CC=C1O
404-86-4
C18H27NO3
305.42
YKPUWZUDDOIDPM-SOFGYWHQSA-N
(6E)-N-[(4-hydroxy-3-methoxyphenyl)methyl]-8-methylnon-6-enamide
COC1=CC(CNC(=O)CCCC\C=C\C(C)C)=CC=C1O

C1869, 404-86-4, C18H27NO3

Spectrum™ Capsaicin, USP is used as an analgesic in nasal spays, topical ointments and in dermal patches to relieve pain. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Loss on Drying 1%
Ignition Residue 1%
Packaging Amber Glass Bottle
Quantity 25 g
Melting Range 57-66°C
CAS Max % 100%
UN Number UN2811