Learn More
Cytiva (Formerly Pall Lab) Mini Profile Capsules - Profile Star Filter, 3 m (3/pkg)
Supplier:  Cytiva 12071
Encompass_Preferred
Scaleable Depth Capsules for Prefiltration and Clarification-High capacity depth medium is absolute particle rated for reliability.-Filter materials offer broad chemical compatibility.-Optimized for viscous fluids. -Mini Profile II filters have a thick depth structure, providing high capacity for larger solids and gels as well as for fine particles.-Profile Star filters feature high area star-shaped pleated construction. The patented design combines the advantages of thick depth filters with those of traditional high area pleated filters. This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-206-3174
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.