Learn More
Molecular Probes™ CaptAvidin™ Biotin-Binding Protein
CaptAvidin™ biotin-binding protein
Supplier: Molecular Probes™ C21385
Description
CaptAvidin™ biotin-binding protein contains a nitrated tyrosine in its biotin-binding site. This chemical modification permits dissociation of the avidin-biotin complex under milder conditions than the traditional avidin-biotin complex. At pH 4.0, CaptAvidin™ biotin-binding protein binds biotin tightly with a Ka of ∼109M-1. This association can then be reversed at pH 10, allowing complete dissociation of the avidin-biotin complex. This is in contrast to the harsh conditions (8 M guanidine hydrochloride, pH 1.5) needed to dissociate the avidin-biotin complex.Specifications
Unconjugated | |
Room Temperature | |
CaptAvidin™ |
Store in freezer (-5 to -30°C). | |
Solid | |
Avidin (unconjugated, reversible biotin binding) |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only. Not for use in diagnostic procedures.