Learn More
Cytiva Capto™ DEAE Chromatography Media
Used for fast, efficient capture and intermediate protein purification
$359.00
Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
45-002-960
View Documents
Cytiva
17544301
100mL
Each for $359.00
Add to cart
Description
Description
  • Composed of high-flow agarose matrix modified with dextran surface extenders and weak diethylaminoethyl (DEAE) anion exchanger
  • Combination of high volume throughput and dynamic binding capacity at high flow
  • Withstands effective CIP procedure and is chemically stable
  • Available in range of different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development