Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Captopril, USP, 98%, Spectrum™

Click to view available options
Quantity:
1 g
5 g
25 g
100 g
500 g
1 kg
Packaging:
Amber Glass Bottle
62571-86-2
C9H15NO3S
217.28
FAKRSMQSSFJEIM-RQJHMYQMSA-N
(2S)-1-[(2S)-2-methyl-3-sulfanylpropanoyl]pyrrolidine-2-carboxylic acid
C[C@H](CS)C(=O)N1CCC[C@H]1C(O)=O
62571-86-2
C9H15NO3S
217.28
FAKRSMQSSFJEIM-RQJHMYQMSA-N
(2S)-1-[(2S)-2-methyl-3-sulfanylpropanoyl]pyrrolidine-2-carboxylic acid
C[C@H](CS)C(=O)N1CCC[C@H]1C(O)=O

C1426, 62571-86-2, C9H15NO3S

Spectrum™ Captopril, USP is an angiotensin-converting enzyme (ACE) inhibitor used for the treatment of hypertension and some types of congestive heart failure. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Packaging Amber Glass Bottle
Quantity 1 kg
CAS Max % 1
Percent Purity 98%