Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD Capture Assay Set
Measurement of cytokines levels in-vivo
$636.50
Specifications
Quantity | 10 Plates |
---|---|
Format | Set |
Includes | Plate coating Antibody, in vivo capture Antibody, IVC standard, Enzyme concentrate, technical data sheet and lot specific certificate of analysis |
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
BDB558491
|
BD
558491 |
Mouse IFN-gamma capture assay set |
Each for $636.50
|
|
BDB558072
|
BD
558072 |
IL-10 Capture assay set | This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
- Reactivity: Mouse
- Format: Set
In-vitro capture assay
Specifications
10 Plates | |
Set | |
Plate coating Antibody, in vivo capture Antibody, IVC standard, Enzyme concentrate, technical data sheet and lot specific certificate of analysis |
Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title