Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CAPZA1 Mouse anti-Human, DyLight™ 405, Clone: OTI2G4, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270336V

Catalog No. NBP270336V


Add to cart

Description

Description

CAPZA1 Monoclonal antibody specifically detects CAPZA1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.
Specifications

Specifications

CAPZA1
OTI2G4
Western Blot, Flow Cytometry, Immunohistochemistry
Affinity Purified, Conjugated
CAPZA1
Full length recombinant protein of human CAPZA1 (NP_006126) produced in HEK293T cells.
Immunogen affinity purified
Extracellular Matrix
Primary
829.0
Flow Cytometry, Immunohistochemistry, Western Blot
DyLight 405
50mM Sodium Borate with 0.05% Sodium Azide
Cap Z, CAPPA1, capping protein (actin filament) muscle Z-line, alpha 1, CAPZ, CapZ alpha-1, CAZ1, F-actin-capping protein subunit alpha-1
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit