Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CAPZA1 Mouse anti-Human, DyLight 650, Clone: OTI2G4, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP270336C
Description
CAPZA1 Monoclonal antibody specifically detects CAPZA1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.Specifications
CAPZA1 | |
OTI2G4 | |
Western Blot, Flow Cytometry, Immunohistochemistry | |
Affinity Purified, Conjugated | |
CAPZA1 | |
Full length recombinant protein of human CAPZA1 (NP_006126) produced in HEK293T cells. | |
Immunogen affinity purified | |
Extracellular Matrix | |
Primary | |
829.0 |
Flow Cytometry, Immunohistochemistry, Western Blot | |
DyLight 650 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Cap Z, CAPPA1, capping protein (actin filament) muscle Z-line, alpha 1, CAPZ, CapZ alpha-1, CAZ1, F-actin-capping protein subunit alpha-1 | |
Mouse | |
IgG1 | |
0.1 mL | |
Store at 4°C in the dark. | |
Monoclonal | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title