Learn More
CAPZA2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 159481AP
Catalog No. 50-173-0591
Add to cart
Description
Description
The protein encoded by this gene is a member of the F-actin capping protein alpha subunit family. It is the alpha subunit of the barbed-end actin binding protein Cap Z. By capping the barbed end of actin filaments, Cap Z regulates the growth of the actin filaments at the barbed end.
Specifications
Specifications
CAPZA2
0.23 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P47754, P47755, Q3T1K5
CAPPA2, CAPZ, CapZ alpha 2, CAPZA2
Rabbit
IgG
150 μL
-20°C
Primary
12343, 493810, 830
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
CAPZA2
Liquid
Capza2
CAPZA2 Fusion Protein Ag8544
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat