Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CAPZA3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA550369
Description
The antibody detects endogenous levels of total CAPZA3 protein.
CAPZA3 Polyclonal antibody specifically detects CAPZA3 in Human samples. It is validated for Immunohistochemistry (Paraffin).Specifications
CAPZA3 | |
Polyclonal | |
Unconjugated | |
CAPZA3 | |
510-4; CAPAA3; Cappa3; capping actin protein of muscle Z-line alpha subunit 3; capping actin protein of muscle Z-line subunit alpha 3; capping protein (actin filament) muscle Z-line, alpha 3; capping protein alpha 3; capZ alpha-3; Capza3; cp alpha3; CP-alpha-3; epididymis secretory protein Li 86; F-actin capping protein alpha-3 subunit; F-actin-capping protein subunit alpha-3; Germ cell-specific protein 3; GSG3; HEL-S-86; repro32; testis tissue sperm-binding protein Li 73n; Tex8 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
93661 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
2.9 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q96KX2 | |
CAPZA3 | |
Fusion protein corresponding to a region derived from internal residues of human capping protein (actin filament) muscle Z-line, alpha 3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction