Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CAPZB Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

Manufacturer:  Proteintech Group IncSupplier Diversity Partner 250431AP

Catalog No. 50-173-4690


Add to cart

Description

Description

CAPZB is a member of the F-actin capping protein family. This gene encodes the beta subunit of the barbed-end actin binding protein. The protein regulates growth of the actin filament by capping the barbed end of growing actin filaments.
Specifications

Specifications

CAPZB
0.51 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P47756, P47757
CAPB, CAPPB, CAPZ, CapZ beta, CAPZB
Rabbit
IgG
150 μL
-20°C
Primary
12345, 832
Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
CAPZB
Liquid
CAPZB
CAPZB Fusion Protein Ag19327
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit