Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CAR/NR1I3 Mouse anti-Human, Alexa Fluor™ 700, Clone: OTI1C6, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271525AF700

Catalog No. N271525A700


Add to cart

Description

Description

CAR/NR1I3 Monoclonal antibody specifically detects CAR/NR1I3 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

CAR/NR1I3
OTI1C6
Western Blot, Immunohistochemistry
Affinity Purified, Conjugated
NR1I3
Recombinant protein fragment corresponding to amino acids 1-236 of human CAR/NR1I3 (NP_001070950) produced in E.coli.
Immunogen affinity purified
Lipid and Metabolism
Primary
9970.0
Immunohistochemistry, Western Blot
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
CARCAR1, Constitutive activator of retinoid response, constitutive active receptor, Constitutive active response, Constitutive androstane receptor, MB67, MGC150433, MGC97144, MGC97209, nuclear receptor subfamily 1 group I member 3, nuclear receptor subfamily 1, group I, member 3, orphan nuclear hormone receptor, Orphan nuclear receptor MB67
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit