Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Caramel Color, Food Grade, Spectrum™

Click to view available options
Quantity:
500 mL
4 L
25 kg
Packaging:
Amber Glass Bottle
Poly Drum
8028-89-5
8028-89-5

C1882, 8028-89-5

Spectrum™ Caramel Color, Food Grade
Quantity 25 kg
Specific Gravity 1.315-1.325
Packaging Poly Drum
CAS Max % 100%