Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Caraway Oil, FCC, 50%, Spectrum™

Click to view available options
Quantity:
25 lbs.
Packaging:
Steel Pail

C1094, 8000-42-8

Spectrum™ Caraway Oil, FCC is used as a food flavoring agent. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Quantity 25 lbs.
Specific Gravity 0.900-0.910
Refractive Index 1.484-1.488
Packaging Steel Pail
CAS 8000-42-8
CAS Max % 100%
UN Number UN1197
Percent Purity 50%