Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Carbacyclin, 99%

Click to view available options
Quantity:
0.5mg
1mg
69552-46-1
C21H34O4
350.499
MFCD00210824
XZFRIPGNUQRGPI-GRIBHNIZSA-N
carboprostacyclin,carbacyclin,carbaprostacyclin,carbacycline,carba pgx,6a-carba-pgi2,6,9-methano pgi2,6,9-methano-pgi2,6,9-methanoprostaglandin i2,5e-6a-carba-prostaglandin i2
133126726
(5E)-5-[(3aR,4S,5S,6aR)-5-hydroxy-4-[(E,3R)-3-hydroxyoct-1-enyl]-3,3a,4,5,6,6a-hexahydro-1H-pentalen-2-ylidene]pentanoic acid
CCCCCC(C=CC1C(CC2C1CC(=CCCCC(=O)O)C2)O)O
69552-46-1
C21H34O4
350.499
MFCD00210824
XZFRIPGNUQRGPI-GRIBHNIZSA-N
carboprostacyclin,carbacyclin,carbaprostacyclin,carbacycline,carba pgx,6a-carba-pgi2,6,9-methano pgi2,6,9-methano-pgi2,6,9-methanoprostaglandin i2,5e-6a-carba-prostaglandin i2
133126726
(5E)-5-[(3aR,4S,5S,6aR)-5-hydroxy-4-[(E,3R)-3-hydroxyoct-1-enyl]-3,3a,4,5,6,6a-hexahydro-1H-pentalen-2-ylidene]pentanoic acid
CCCCCC(C=CC1C(CC2C1CC(=CCCCC(=O)O)C2)O)O
Quantity 1mg
Melting Point 65°C to 67°C
Color White
Formula Weight 350.5
Physical Form Solid
Percent Purity 99%
Chemical Name or Material Carbacyclin

storageNote1 : -30° to -10°C