Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carbenicillin disodium salt, 50mg/mL in distilled water, sterile-filtered, Alfa Aesar™

Click to view available options
Quantity:
50mL
100mL
4800-94-6
C17H16N2Na2O6S
422.363
MFCD00077683
RTYJTGSCYUUYAL-YCAHSCEMSA-L
20933
CHEBI:34609
disodium;(2S,5R,6R)-6-[(2-carboxylato-2-phenylacetyl)amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
CC1(C(N2C(S1)C(C2=O)NC(=O)C(C3=CC=CC=C3)C(=O)[O-])C(=O)[O-])C.[Na+].[Na+]
4800-94-6
C17H16N2Na2O6S
422.363
MFCD00077683
RTYJTGSCYUUYAL-YCAHSCEMSA-L
20933
CHEBI:34609
disodium;(2S,5R,6R)-6-[(2-carboxylato-2-phenylacetyl)amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
CC1(C(N2C(S1)C(C2=O)NC(=O)C(C3=CC=CC=C3)C(=O)[O-])C(=O)[O-])C.[Na+].[Na+]

Used for selection of ampr transformed cells and to study the role of penicillin-sensitive transpeptidases in cell wall biosynthesis.

Applications
Used for selection of ampr transformed cells and to study the role of penicillin-sensitive transpeptidases in cell wall biosynthesis. Carboxypenicillin antibiotic that inhibits bacterial cell-wall synthesis (peptidoglycan cross-linking) by inactivating transpeptidases on the inner surface of the bacterial cell membrane. Analog to ampicillin, Antimicrobial spectrum.

Solubility
Soluble in water.

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
Physical Form Liquid
Concentration 50mg/mL in distilled water
Quantity 50mL
Chemical Name or Material Carbenicillin disodium salt, sterile-filtered

GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.

GHS P Statement
P285-P261-P280-P342+P311-P363-P501a
In case of inadequate ventilation wear respiratory protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wash contaminated clothing before reuse.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : ON9105000

storageNote1 : −30° to −10°C